accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HNON

CAS RN 603-51-0

IUPAC Name: 3-amino-1,1-diphenylurea
Synonyms: 3-amino-1,1-diphenylurea
Molecular Weight (g/mol): 227.27
Molecular Formula: C13H13N3O
InChi Key: VVVFQQJJJRFDTE-UHFFFAOYSA-N
SMILES: NNC(=O)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results