Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Diphenylsemicarbazide 98%, Thermo Scientific™

Catalog No. AC199920250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC199920250 25 g
AC199920050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC199920250 Supplier Thermo Scientific Chemicals Supplier No. 199920250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-51-0
Molecular Formula C13H13N3O
Molecular Weight (g/mol) 227.27
MDL Number MFCD00007592
InChI Key VVVFQQJJJRFDTE-UHFFFAOYSA-N
Synonym 4,4-diphenylsemicarbazide, n,n-diphenylhydrazinecarboxamide, hydrazinecarboxamide, n,n-diphenyl, semicarbazide, 4,4-diphenyl, qa7tgp7xir, unii-qa7tgp7xir, hydrazinecarboxamide,n,n-diphenyl, semicarbazide,4-diphenyl, hydrazinecarboxamide,n-diphenyl, 4,4-diphenylsemicar-bazide
PubChem CID 69049
IUPAC Name 3-amino-1,1-diphenylurea
SMILES NNC(=O)N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 147°C to 151°C
Color White to Brown
Infrared Spectrum Authentic
Linear Formula (C6H5)2NCONHNH2
Quantity 25 g
Beilstein 12, I, 257
Formula Weight 227.27
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4-Diphenylsemicarbazide, 98%
Show More Show Less
EINECSNumber 210-046-5
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.