accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(S)O(R)OH(S)HO(S)HO(R)OH

CAS RN 6032-32-2

IUPAC Name: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol
Synonyms: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol): 286.28
Molecular Formula: C13H18O7
InChi Key: SIXFVXJMCGPTRB-UJPOAAIJSA-N
SMILES: COC1=CC=C(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)C=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)