Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenyl beta-D-Glucopyranoside 97.0+%, TCI America™
SDP

Catalog No. M16315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16315G 5 g
M163125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16315G Supplier TCI America Supplier No. M16315G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6032-32-2 | C13H18O7 | 286.28 g/mol

Chemical Identifiers

CAS 6032-32-2
Molecular Formula C13H18O7
Molecular Weight (g/mol) 286.28
MDL Number MFCD06797143
InChI Key SIXFVXJMCGPTRB-UJPOAAIJSA-N
Synonym methylarbutin, 4-methoxyphenyl beta-d-glucopyranoside, p-methoxyphenyl b-d-glucoside, unii-f9mw5fu0xh, 4-methoxyphenyl b-d-glucopyranoside, f9mw5fu0xh, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-4-methoxyphenoxy oxane-3,4,5-triol, methylarbutoside, 4-methoxyphenylglucoside, p-methoxyphenyl beta-d-glucopyranoside
PubChem CID 80131
ChEBI CHEBI:81207
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 176°C
Color White
Quantity 5 g
Formula Weight 286.28
Percent Purity ≥97.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl beta-D-Glucopyranoside
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.