accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60345-97-3

OHOO

CAS RN 60345-97-3

IUPAC Name: 3-(2-hydroxyethoxy)benzaldehyde
Synonyms: 3-(2-hydroxyethoxy)benzaldehyde
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: LIUCHRXQRHVSJI-UHFFFAOYSA-N
SMILES: OCCOC1=CC=CC(C=O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)