Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Hydroxyethoxy)benzaldehyde 96.0+%, TCI America™
SDP

Catalog No. H08585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08585G 5 g
H085825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08585G Supplier TCI America Supplier No. H08585G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60345-97-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00191448
InChI Key LIUCHRXQRHVSJI-UHFFFAOYSA-N
PubChem CID 6453965
IUPAC Name 3-(2-hydroxyethoxy)benzaldehyde
SMILES OCCOC1=CC=CC(C=O)=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(2-Hydroxyethoxy)benzaldehyde
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.