accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNHO

CAS RN 60399-02-2

IUPAC Name: 2-[(methylamino)methyl]phenol
Synonyms: 2-[(methylamino)methyl]phenol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: PVFSNBPKQDSTPP-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results