accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3H3CH3COCH3O

CAS RN 604-35-3

IUPAC Name: 9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl acetate
Synonyms: cholesteryl acetate
Molecular Weight (g/mol): 428.7
Molecular Formula: C29H48O2
InChi Key: XUGISPSHIFXEHZ-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)C1CCC2C3CC=C4CC(CCC4(C)C3CCC12C)OC(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)