Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesteryl acetate, 97+%, Thermo Scientific Chemicals

Catalog No. AAA1505214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1505214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1505214 Supplier Thermo Scientific Chemicals Supplier No. A1505214

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Cholesteryl acetate is used in cosmetics, wrist watches, thermometers, propane tank volume indicators, video displays, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cholesteryl acetate is used in cosmetics, wrist watches, thermometers, propane tank volume indicators, video displays, pharmaceuticals.

Solubility
Soluble in chloroform, ethanol or ethyl acetate.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-35-3
Molecular Formula C29H48O2
Molecular Weight (g/mol) 428.70
MDL Number MFCD00003636,MFCD00003636,MFCD00003636
InChI Key XUGISPSHIFXEHZ-UHFFFAOYNA-N
Synonym cholesteryl acetate
PubChem CID 12360660
SMILES CC(C)CCCC(C)C1CCC2C3CC=C4CC(CCC4(C)C3CCC12C)OC(C)=O

Specifications

Melting Point 112°C to 114°C
Quantity 25 g
Beilstein 2064235
Merck Index 14,2201
Solubility Information Soluble in chloroform,ethanol or ethyl acetate.
Optical Rotation −43° (c=5 in Chloroform)
IUPAC Name [(10R,13R)-10,13-dimethyl-17-(6-methylheptan-2-yl)-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
Formula Weight 428.7
Percent Purity ≥97%
Chemical Name or Material Cholesteryl acetate
EINECSNumber 210-066-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.