accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60444-78-2

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 4-formylbenzoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 4-formylbenzoate
Molecular Weight (g/mol): 247.21
Molecular Formula: C12H9NO5
InChi Key: VHYRHFNOWKMCHQ-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)C(=O)ON1C(=O)CCC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results