Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl 4-Formylbenzoate 98.0+%, TCI America™
SDP

Catalog No. S0893100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0893100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0893100MG Supplier TCI America Supplier No. S0893100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60444-78-2
Molecular Formula C12H9NO5
Molecular Weight (g/mol) 247.206
MDL Number MFCD00133733
InChI Key VHYRHFNOWKMCHQ-UHFFFAOYSA-N
Synonym 4-Formylbenzoic Acid N-Succinimidyl Ester
PubChem CID 4126130
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-formylbenzoate
SMILES C1CC(=O)N(C1=O)OC(=O)C2=CC=C(C=C2)C=O

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 100 mg
Formula Weight 247.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 4-Formylbenzoate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.