accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6048-68-6

IUPAC Name: 2,5,8,11,14,17,20,23,26-nonaoxaoctacosan-28-ol
Synonyms: O-methyl-nonaethyleneglycol
Molecular Weight (g/mol): 428.52
Molecular Formula: C19H40O10
InChi Key: VVHAVLIDQNWEKF-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCOCCOCCOCCOCCOCCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results