Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonaethylene Glycol Monomethyl Ether 93.0+%, TCI America™
SDP

Catalog No. N0699500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0699500MG 500 mg
N06991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0699500MG Supplier TCI America Supplier No. N0699500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6048-68-6
Molecular Formula C19H40O10
Molecular Weight (g/mol) 428.52
MDL Number MFCD06797148
InChI Key VVHAVLIDQNWEKF-UHFFFAOYSA-N
Synonym mPEG9-Alcohol
PubChem CID 11339376
ChEBI CHEBI:59168
IUPAC Name 2,5,8,11,14,17,20,23,26-nonaoxaoctacosan-28-ol
SMILES COCCOCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

Melting Point 14°C
Color Yellow
Quantity 500 mg
Formula Weight 428.52
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonaethylene Glycol Monomethyl Ether
EINECSNumber (7)-0097
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.