accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 605-32-3

IUPAC Name: 2-hydroxy-9,10-dihydroanthracene-9,10-dione
Synonyms: 2-hydroxyanthraquinone 2-hydroxyanthra-9,10-quinone
Molecular Weight (g/mol): 224.22
Molecular Formula: C14H8O3
InChi Key: GCDBEYOJCZLKMC-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results