Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyanthraquinone 97.0+%, TCI America™
SDP

Catalog No. H1012250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1012250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H1012250G Supplier TCI America Supplier No. H1012250G

Specifications

CAS 605-32-3
Molecular Formula C14H8O3
MDL Number MFCD00027374
Quantity 250 g
InChI Key GCDBEYOJCZLKMC-UHFFFAOYSA-N
SMILES OC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
IUPAC Name 2-hydroxy-9,10-dihydroanthracene-9,10-dione
Molecular Weight (g/mol) 224.22
PubChem CID 11796
ChEBI CHEBI:37482
Formula Weight 224.22
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxyanthraquinone
Show More Show Less
RTECSNumber CB7250000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.