accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNOSOOHOCH3NHNOSOOHONaNa(E/Z)(E/Z)

CAS RN 6059-34-3

IUPAC Name: 3-(2-{3,3'-dimethyl-4'-[2-(1-oxo-4-sulfo-1,2-dihydronaphthalen-2-ylidene)hydrazin-1-yl]-[1,1'-biphenyl]-4-yl}hydrazin-1-ylidene)-4-oxo-3,4-dihydronaphthalene-1-sulfonic acid disodium
Synonyms: 3-(2-{3,3'-dimethyl-4'-[2-(1-oxo-4-sulfonaphthalen-2-ylidene)hydrazin-1-yl]-[1,1'-biphenyl]-4-yl}hydrazin-1-ylidene)-4-oxonaphthalene-1-sulfonic acid disodium
Molecular Weight (g/mol): 728.7
Molecular Formula: C34H26N4Na2O8S2
InChi Key: UIHNTQIDLGWLFG-UHFFFAOYSA-N
SMILES: [Na].[Na].CC1=C(NN=C2C=C(C3=CC=CC=C3C2=O)S(O)(=O)=O)C=CC(=C1)C1=CC(C)=C(NN=C2C=C(C3=CC=CC=C3C2=O)S(O)(=O)=O)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results