Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azo Blue, TCI America™
SDP

Catalog No. A058725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A058725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A058725G Supplier TCI America Supplier No. A058725G

Chemical Identifiers

CAS 6059-34-3
Molecular Formula C34H26N4Na2O8S2
Molecular Weight (g/mol) 728.70
MDL Number MFCD00059133
InChI Key UIHNTQIDLGWLFG-UHFFFAOYSA-N
Synonym Direct Violet 39
PubChem CID 131864345
IUPAC Name 3-(2-{3,3'-dimethyl-4'-[2-(1-oxo-4-sulfo-1,2-dihydronaphthalen-2-ylidene)hydrazin-1-yl]-[1,1'-biphenyl]-4-yl}hydrazin-1-ylidene)-4-oxo-3,4-dihydronaphthalene-1-sulfonic acid disodium
SMILES [Na].[Na].CC1=C(NN=C2C=C(C3=CC=CC=C3C2=O)S(O)(=O)=O)C=CC(=C1)C1=CC(C)=C(NN=C2C=C(C3=CC=CC=C3C2=O)S(O)(=O)=O)C=C1

Specifications

Color Black
Quantity 25 g
Formula Weight 726.69
Physical Form Crystalline Powder
Chemical Name or Material Azo Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.