accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 606-00-8

IUPAC Name: methyl 2-amino-3,5-dibromobenzoate
Synonyms: methyl 2-amino-3,5-dibromobenzoate
Molecular Weight (g/mol): 308.96
Molecular Formula: C8H7Br2NO2
InChi Key: NGXVMFCGYYHEGC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=CC(Br)=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results