Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,5-Dibromoanthranilate 98.0+%, TCI America™
SDP

Catalog No. M188625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M188625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M188625G Supplier TCI America Supplier No. M188625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 606-00-8
Molecular Formula C8H7Br2NO2
Molecular Weight (g/mol) 308.96
MDL Number MFCD00010873
InChI Key NGXVMFCGYYHEGC-UHFFFAOYSA-N
Synonym methyl 3,5-dibromoanthranilate, benzoic acid, 2-amino-3,5-dibromo-, methyl ester, 3,5-dibromoanthranilic acid methyl ester, 2-amino-3,5-dibromobenzoic acid methyl ester, anthranilic acid, 3,5-dibromo-, methyl ester, methyl 2-amino-3,5-dibromo-benzoate, pubchem3352, acmc-209mkw, ksc490q2l, methyl,3,5-dibromoanthranilate
PubChem CID 69069
IUPAC Name methyl 2-amino-3,5-dibromobenzoate
SMILES COC(=O)C1=CC(Br)=CC(Br)=C1N

Specifications

Melting Point 88°C
Quantity 25 g
Formula Weight 308.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,5-Dibromoanthranilate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.