accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 606-20-2

IUPAC Name: 2-methyl-1,3-dinitrobenzene
Synonyms: 2,6-dinitrotoluene 2,6-dinitromethylbenzene
Molecular Weight (g/mol): 182.14
Molecular Formula: C7H6N2O4
InChi Key: XTRDKALNCIHHNI-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)