Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dinitrotoluene 99.0+%, TCI America™
SDP

Catalog No. D115125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D115125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D115125G Supplier TCI America Supplier No. D115125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 606-20-2
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.135
MDL Number MFCD00007158
InChI Key XTRDKALNCIHHNI-UHFFFAOYSA-N
Synonym 2,6-dinitrotoluene, 2,6-dnt, benzene, 2-methyl-1,3-dinitro, 1-methyl-2,6-dinitrobenzene, 2,6-dinitromethylbenzene, toluene, 2,6-dinitro, rcra waste number u106, 2-methyl-1,3-dinitro-benzene, unii-gg7fav92mk, ccris 1006
PubChem CID 11813
ChEBI CHEBI:957
IUPAC Name 2-methyl-1,3-dinitrobenzene
SMILES CC1=C(C=CC=C1[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 65°C
Color Yellow
Quantity 25 g
UN Number 3454
Formula Weight 182.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dinitrotoluene
EINECSNumber (3)-0446
RTECSNumber XT1925000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.