accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 606-23-5

IUPAC Name: 2,3-dihydro-1H-indene-1,3-dione
Synonyms: 1,3-indandione
Molecular Weight (g/mol): 146.15
Molecular Formula: C9H6O2
InChi Key: UHKAJLSKXBADFT-UHFFFAOYSA-N
SMILES: O=C1CC(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (1)
  • (7)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (6)
  • (1)
  • (4)
  • (3)
  • (1)
  • (7)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)