accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60628-96-8

IUPAC Name: 1-({[1,1'-biphenyl]-4-yl}(phenyl)methyl)-1H-imidazole
Synonyms: bifonazole bifokey dermapharm brand of bifonazole bioglan brand of bifonazole mycospor bifonazole (substance) 联苯苄唑 bifomyk merck lipha santé brand of bifonazole amycor
Molecular Weight (g/mol): 310.4
Molecular Formula: C22H18N2
InChi Key: OCAPBUJLXMYKEJ-UHFFFAOYNA-N
SMILES: C1=CN(C=N1)C(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results