Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bifonazole, 98%, Thermo Scientific™

Catalog No. AC458620250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458620250 25 g
AC458620010 1 g
AC458620050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC458620250 Supplier Thermo Scientific Chemicals Supplier No. 458620250
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60628-96-8
Molecular Formula C22H18N2
Molecular Weight (g/mol) 310.39
InChI Key OCAPBUJLXMYKEJ-UHFFFAOYSA-N
Synonym bifonazole, mycospor, trifonazole, bifonazol, bifonazolum, amycor, azolmen, bifonazol inn-spanish, bifonazolum inn-latin, 1-4-biphenylyl phenylmethyl-1h-imidazole
PubChem CID 2378
ChEBI CHEBI:78692
IUPAC Name 1-[phenyl-(4-phenylphenyl)methyl]imidazole
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4

Specifications

Melting Point 149.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 1 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 310.39
Percent Purity 98%
Chemical Name or Material Bifonazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 262-336-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.