accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60633-25-2

IUPAC Name: 2-bromo-4-chloro-1-methoxybenzene
Synonyms: 2-bromo-4-chloro-1-methoxybenzene
Molecular Weight (g/mol): 221.48
Molecular Formula: C7H6BrClO
InChi Key: YJEMGEBDXDPBSP-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C=C1Br
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)