Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-chloroanisole 98.0+%, TCI America™
SDP

Catalog No. B31115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31115G 5 g
B311125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31115G Supplier TCI America Supplier No. B31115G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60633-25-2
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.478
MDL Number MFCD00079705
InChI Key YJEMGEBDXDPBSP-UHFFFAOYSA-N
Synonym 2-bromo-4-chloroanisole, 2-bromo-4-chloro-1-methoxy-benzene, benzene, 2-bromo-4-chloro-1-methoxy, 4-chloro-2-bromoanisole, pubchem2629, 2-bromo4-chloroanisole, 4-chloro-2-bromanisole, 2-bromo-4-chloro anisole, acmc-1b9j6, ksc490k3r
PubChem CID 521935
IUPAC Name 2-bromo-4-chloro-1-methoxybenzene
SMILES COC1=C(C=C(C=C1)Cl)Br

Specifications

Color Yellow
Boiling Point 127°C
Quantity 5 g
Formula Weight 221.48
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-chloroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.