accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NCH3H3CNCH3CH3H2OH2OH2OH2OH2OH2OH2OH2OH2OCl

CAS RN 60662-33-1

IUPAC Name: 4-{bis[4-(dimethylamino)phenyl]methylidene}-N,N-dimethylcyclohexa-2,5-dien-1-iminium nonahydrate chloride
Synonyms: gentian violet nonahydrate chloride
Molecular Weight (g/mol): 570.12
Molecular Formula: C25H48ClN3O9
InChi Key: QCYZZDDACAOKCV-UHFFFAOYSA-M
SMILES: O.O.O.O.O.O.O.O.O.[Cl-].CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)=C1C=CC(C=C1)=[N+](C)C
Color 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results