Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal Violet Nonahydrate 97.0+%, TCI America™
SDP

Catalog No. A51041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A51041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A51041G Supplier TCI America Supplier No. A51041G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ion association reagent for spectrophotometric analysis.

Chemical Identifiers

CAS 60662-33-1 , 548-62-9
Molecular Formula C25H48ClN3O9
Molecular Weight (g/mol) 570.121
MDL Number MFCD00011750
InChI Key QCYZZDDACAOKCV-UHFFFAOYSA-M
Synonym Basic Violet 3
PubChem CID 91658927
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride;nonahydrate
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.O.O.O.O.O.O.O.O.O.[Cl-]

Specifications

Melting Point 211°C
UN Number 3077
Color Green/Yellow
Quantity 1 g
Physical Form Crystal/Powder
Formula Weight 407.99
Percent Purity ≥97.0% (T)
Grade Spectrophotometry
Chemical Name or Material Crystal Violet Nonahydrate [Ion association reagent for spectrophotometric analysis]
EINECSNumber (5)-1971&(4)-0873
RTECSNumber BO9000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.