accessibility menu, dialog, popup

UserName

Viewing profile as user:
BH3CCH3OSOOFFF

CAS RN 60669-69-4

IUPAC Name: dibutylboranyl trifluoromethanesulfonate
Synonyms: dibutylboron trifluoromethanesulfonate
Molecular Weight (g/mol): 274.11
Molecular Formula: C9H18BF3O3S
InChi Key: FAVAVMFXAKZTMV-UHFFFAOYSA-N
SMILES: CCCCB(CCCC)OS(=O)(=O)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results