accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6068-62-8

IUPAC Name: (2E)-1,1,4,4-tetramethoxybut-2-ene
Synonyms: 1,1,4,4-tetramethoxy-but-2-ene
Molecular Weight (g/mol): 176.21
Molecular Formula: C8H16O4
InChi Key: ZFGVCDSFRAMNMT-AATRIKPKSA-N
SMILES: COC(OC)\C=C\C(OC)OC
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (4)