accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6069-98-3

IUPAC Name: 1-methyl-4-(propan-2-yl)cyclohexane
Synonyms: p-menthane p-menthane skeleton
Molecular Weight (g/mol): 140.27
Molecular Formula: C10H20
InChi Key: CFJYNSNXFXLKNS-UHFFFAOYSA-N
SMILES: CC(C)C1CCC(C)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results