Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1-Isopropyl-4-methylcyclohexane, TCI America™
SDP

Catalog No. I02845ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02845ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I02845ML Supplier TCI America Supplier No. I02845ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6069-98-3
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00070477
InChI Key CFJYNSNXFXLKNS-UHFFFAOYSA-N
Synonym cis-Hexahydro-p-cymene
PubChem CID 7459
ChEBI CHEBI:25826
IUPAC Name 1-methyl-4-propan-2-ylcyclohexane
SMILES CC1CCC(CC1)C(C)C

Specifications

Color Colorless
Boiling Point 167°C
Quantity 5 mL
UN Number 3295
Formula Weight 140.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-1-Isopropyl-4-methylcyclohexane
EINECSNumber (3)-2246
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.