CAS RN 607-24-9
IUPAC Name:
2-nitronaphthalen-1-ol
Synonyms:
2-nitronaphthalen-1-ol
Molecular Weight (g/mol):
189.17
Molecular Formula:
C10H7NO3
InChi Key:
MUCCHGOWMZTLHK-UHFFFAOYSA-N
SMILES:
OC1=C(C=CC2=CC=CC=C12)[N+]([O-])=O
1
–
2
of
2
results
1
Sigma Aldrich 2-Nitro-1-naphthol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.