accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 607-32-9

IUPAC Name: 5-nitroisoquinoline
Synonyms: 5-nitro isoquinoline
Molecular Weight (g/mol): 174.16
Molecular Formula: C9H6N2O2
InChi Key: PYGMPFQCCWBTJQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2C=CN=CC2=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)