Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitroisoquinoline, TCI America™
SDP

Catalog No. N06405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06405G Supplier TCI America Supplier No. N06405G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 607-32-9
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD00006905
InChI Key PYGMPFQCCWBTJQ-UHFFFAOYSA-N
Synonym isoquinoline, 5-nitro, 5-nitro-isoquinoline, 5-nitro isoquinoline, 5-nitroisochinolin, pubchem6272, maybridge1_001820, acmc-1av1q, 5-nitroisoquinoline, pygmpfqccwbtjq-uhfffaoysa
PubChem CID 69085
IUPAC Name 5-nitroisoquinoline
SMILES C1=CC2=C(C=CN=C2)C(=C1)[N+](=O)[O-]

Specifications

Melting Point 108°C
Quantity 5 g
Formula Weight 174.16
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroisoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.