CAS RN 607-66-9
CAS RN 607-66-9
IUPAC Name:
4-methyl-1,2-dihydroquinolin-2-one
Synonyms:
4-methyl-2(1H)-quinolinone 2-lepidinol 2-hydroxylepidine 2(1H)-lepidinone
Molecular Weight (g/mol):
159.19
Molecular Formula:
C10H9NO
InChi Key:
APLVPBUBDFWWAD-UHFFFAOYSA-N
SMILES:
CC1=CC(=O)NC2=CC=CC=C12
1
–
1
of
1
results
1
4-Methylcarbostyril 98.0+%, TCI America™

CAS: 607-66-9 Molecular Formula: C10H9NO Molecular Weight (g/mol): 159.19 MDL Number: MFCD00006745 InChI Key: APLVPBUBDFWWAD-UHFFFAOYSA-N Synonym: 4-methylquinolin-2-ol,2-hydroxy-4-methylquinoline,4-methyl-2-quinolinol,2-hydroxylepidine,4-methylquinolin-2 1h-one,4-methyl-2-quinolone,4-methylcarbostyril,2 1h-lepidinone,2-lepidinol,4-methyl-2 1h-quinolinone PubChem CID: 69088 ChEBI: CHEBI:48986 IUPAC Name: 4-methyl-1,2-dihydroquinolin-2-one SMILES: CC1=CC(=O)NC2=CC=CC=C12