Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylcarbostyril 98.0+%, TCI America™
SDP

Catalog No. H025925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H025925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H025925G Supplier TCI America Supplier No. H025925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity organic building block for research applications.

Chemical Identifiers

CAS 607-66-9
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00006745
InChI Key APLVPBUBDFWWAD-UHFFFAOYSA-N
Synonym 4-methylquinolin-2-ol, 2-hydroxy-4-methylquinoline, 4-methyl-2-quinolinol, 2-hydroxylepidine, 4-methylquinolin-2 1h-one, 4-methyl-2-quinolone, 4-methylcarbostyril, 2 1h-lepidinone, 2-lepidinol, 4-methyl-2 1h-quinolinone
PubChem CID 69088
ChEBI CHEBI:48986
IUPAC Name 4-methyl-1,2-dihydroquinolin-2-one
SMILES CC1=CC(=O)NC2=CC=CC=C12

Specifications

Melting Point 227°C
Color White
Quantity 25 g
Formula Weight 159.19
Percent Purity ≥98.0% (HPLC,N)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylcarbostyril
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.