accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3

CAS RN 607-86-3

IUPAC Name: 2-methylbenzoyl 2-methylbenzoate
Synonyms: 2-methylbenzoyl 2-methylbenzoate
Molecular Weight (g/mol): 254.29
Molecular Formula: C16H14O3
InChi Key: YLBSXJWDERHYFY-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C(=O)OC(=O)C1=CC=CC=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results