Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbenzoic Anhydride 98.0+%, TCI America™
SDP

Catalog No. M29971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29971G 1 g
M29975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29971G Supplier TCI America Supplier No. M29971G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 607-86-3
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.29
MDL Number MFCD00048077
InChI Key YLBSXJWDERHYFY-UHFFFAOYSA-N
PubChem CID 136405
IUPAC Name 2-methylbenzoyl 2-methylbenzoate
SMILES CC1=CC=CC=C1C(=O)OC(=O)C1=CC=CC=C1C

Specifications

Melting Point 39°C
Color White
Boiling Point 221°C
Quantity 1 g
Formula Weight 254.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylbenzoic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.