accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHNHNNOO(E)

CAS RN 607-96-5

IUPAC Name: (4E)-3-hydroxy-5-methyl-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Synonyms: (4E)-3-hydroxy-5-methyl-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 273.25
Molecular Formula: C13H11N3O4
InChi Key: DJQKTCCAPLEDRJ-FYWRMAATSA-N
SMILES: CC1=CC(=O)C=C(O)\C1=N\NC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results