accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60789-54-0

IUPAC Name: [(4-bromobutoxy)methyl]benzene
Synonyms: [(4-bromobutoxy)methyl]benzene
Molecular Weight (g/mol): 243.14
Molecular Formula: C11H15BrO
InChi Key: ZQRVGYAMRJVUAK-UHFFFAOYSA-N
SMILES: BrCCCCOCC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)