Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 4-Bromobutyl Ether 96.0+%, TCI America™
SDP

Catalog No. B28025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28025G 5 g
B280225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28025G Supplier TCI America Supplier No. B28025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60789-54-0
Molecular Formula C11H15BrO
Molecular Weight (g/mol) 243.144
InChI Key ZQRVGYAMRJVUAK-UHFFFAOYSA-N
Synonym 4-Benzyloxybutyl Bromide, (4-Bromobutoxymethyl)benzene
PubChem CID 4422181
IUPAC Name 4-bromobutoxymethylbenzene
SMILES C1=CC=C(C=C1)COCCCCBr

Specifications

Color Yellow
Boiling Point 115°C
Quantity 5 g
Formula Weight 243.14
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl 4-Bromobutyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.