accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHBrBrBrBrBr

CAS RN 608-71-9

IUPAC Name: pentabromophenol
Synonyms: pentabromophenol C6hbr5O
Molecular Weight (g/mol): 488.59
Molecular Formula: C6HBr5O
InChi Key: SVHOVVJFOWGYJO-UHFFFAOYSA-N
SMILES: OC1=C(Br)C(Br)=C(Br)C(Br)=C1Br
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results