Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentabromophenol 99%, Thermo Scientific™

Catalog No. AC129785000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC129785000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC129785000 Supplier Thermo Scientific Chemicals Supplier No. 129785000
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-71-9
Molecular Formula C6HBr5O
Molecular Weight (g/mol) 488.593
MDL Number MFCD00002147
InChI Key SVHOVVJFOWGYJO-UHFFFAOYSA-N
Synonym pentabromophenol, phenol, pentabromo, flammex 5bp, pentabromfenol, pentabromphenol, pentabromfenol czech, unii-v89a88nmmx, ccris 4853, v89a88nmmx, phenol,2,3,4,5,6-pentabromo
PubChem CID 11852
IUPAC Name 2,3,4,5,6-pentabromophenol
SMILES C1(=C(C(=C(C(=C1Br)Br)Br)Br)Br)O

Specifications

Melting Point 223°C to 228°C
Color Beige-Brown to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula C6Br5OH
Quantity 500 g
Beilstein 06, 206
Formula Weight 488.58
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material Pentabromophenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-167-3
RTECSNumber SM6125000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.