accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3O(S)(S)OCH3O(R)NH3COHH

CAS RN 6080-33-7

IUPAC Name: hydrogen (1R,9S,10S)-3-hydroxy-4,12-dimethoxy-17-methyl-17-azatetracyclo[7.5.3.0¹,¹⁰.0²,⁷]heptadeca-2,4,6,11-tetraen-13-one chloride
Synonyms: sinomenine
Molecular Weight (g/mol): 365.85
Molecular Formula: C19H24ClNO4
InChi Key: YMEVIMJAUHZFMW-VUIDNZEBSA-N
SMILES: [H+].[Cl-].COC1=C[C@@H]2[C@@H]3CC4=CC=C(OC)C(O)=C4[C@]2(CCN3C)CC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results