Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sinomenine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. S09261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S09261G Supplier TCI America Supplier No. S09261G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Specifications

CAS 6080-33-7
Melting Point 233°C
Color White
Molecular Formula C19H24ClNO4
Quantity 1 g
MDL Number MFCD01714307
Synonym sinomenine hydrochloride, sinomenine hcl, cucoline, hydrochloride, unii-2j34hrj45s, sinomeinne hydrochloride, c19h23no4.hcl, 7,8-didehydro-3,7-dimethoxy-4-hydroxy-17-methyl-9-alpha,13-alpha,14-alpha-morphinan-6-one hcl, 9-alpha,13-alpha,14-alpha-morphinan-6-one, 7,8-didehydro-3,7-dimethoxy-4-hydroxy-17-methyl-, hydrochloride, sinomenine hydrocloride, sinomenine-hydrochloride
InChI Key YMEVIMJAUHZFMW-VUIDNZEBSA-N
SMILES CN1CCC23CC(=O)C(=CC2C1CC4=C3C(=C(C=C4)OC)O)OC.Cl
Molecular Weight (g/mol) 365.854
PubChem CID 5464452
Formula Weight 365.85
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sinomenine Hydrochloride
Show More Show Less
RTECSNumber QD2163000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.