accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60835-72-5

IUPAC Name: 15-bromo-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine
Synonyms: 15-bromo-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine
Molecular Weight (g/mol): 347.21
Molecular Formula: C14H19BrO5
InChi Key: GPKJNSIFVWMEEI-UHFFFAOYSA-N
SMILES: BrC1=CC=C2OCCOCCOCCOCCOC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results