accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOO

CAS RN 60835-73-6

IUPAC Name: 2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine-15-carbaldehyde
Synonyms: 2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine-15-carbaldehyde
Molecular Weight (g/mol): 296.32
Molecular Formula: C15H20O6
InChi Key: MBJIKIAWNPEHOR-UHFFFAOYSA-N
SMILES: O=CC1=CC=C2OCCOCCOCCOCCOC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)