accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6084-17-9

IUPAC Name: 2-chloro-1-phenylpropan-1-one
Synonyms: 2-chloro-1-phenylpropan-1-one
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: AXCPQHPNAZONTH-UHFFFAOYNA-N
SMILES: CC(Cl)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results