Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloropropiophenone 95.0+%, TCI America™
SDP

Catalog No. C16475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16475G 5 g
C164725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16475G Supplier TCI America Supplier No. C16475G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6084-17-9
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD03844782
InChI Key AXCPQHPNAZONTH-UHFFFAOYNA-N
Synonym 2-Chloro-1-phenyl-1-propanone
PubChem CID 221281
IUPAC Name 2-chloro-1-phenylpropan-1-one
SMILES CC(Cl)C(=O)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 168.62
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloropropiophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.